missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-threonine Dihydrate, TCI America™

Catalog No. G01431G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01431G Supplier TCI America Supplier No. G01431G

Chemical Identifiers

CAS 7093-70-1
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.17
MDL Number MFCD00038176
InChI Key OLIFSFOFKGKIRH-WUJLRWPWSA-N
Synonym H-Gly-Thr-OH
PubChem CID 111257
ChEBI CHEBI:73920
IUPAC Name (2S,3R)-2-(2-aminoacetamido)-3-hydroxybutanoic acid
SMILES C[C@@H](O)[C@H](NC(=O)CN)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 176.17
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-threonine Dihydrate