accessibility menu, dialog, popup

UserName

CAS RN 70755-47-4

BrH3CNH3CCH3CH3

CAS RN 70755-47-4

IUPAC Name: dihexadecyldimethylazanium bromide
Synonyms: dihexadecyldimethylazanium bromide
Molecular Weight (g/mol): 574.86
Molecular Formula: C34H72BrN
InChi Key: VIXPKJNAOIWFMW-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results