missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyldipalmitylammonium Bromide 97.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 574.86 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dimethyldipalmitylammonium Bromide |
Safety and Handling
| EINECSNumber | (2)-0184&(9)-1971 |
| TSCA | No |