CAS RN 7073-94-1
IUPAC Name:
1-bromo-2-(propan-2-yl)benzene
Synonyms:
1-bromo-2-isopropylbenzene
Molecular Weight (g/mol):
199.09
Molecular Formula:
C9H11Br
InChi Key:
LECYCYNAEJDSIL-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC=C1Br
1
–
1
of
1
results
1
2-Bromocumene 97.0+%, TCI America™
CAS: 7073-94-1 Molecular Formula: C9H11Br Molecular Weight (g/mol): 199.091 MDL Number: MFCD00051567 InChI Key: LECYCYNAEJDSIL-UHFFFAOYSA-N Synonym: 1-bromo-2-isopropylbenzene,1-bromo-2-1-methylethyl benzene,2-isopropylbromobenzene,o-bromocumene,2-bromocumene,2-bromo cumene,cumene, o-bromo,benzene, 1-bromo-2-1-methylethyl,1-bromo-2-propan-2-yl benzene,1-bromo-2-isopropyl benzene PubChem CID: 23475 IUPAC Name: 1-bromo-2-propan-2-ylbenzene SMILES: CC(C)C1=CC=CC=C1Br