missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromocumene 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 210°C |
| Quantity | 5 g |
| Formula Weight | 199.09 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Bromocumene |
Safety and Handling
| TSCA | No |