CAS RN 70681-41-3
CAS RN 70681-41-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethyl D-(+)-Malate 98.0+%, TCI America™
CAS: 70681-41-3 Molecular Formula: C6H10O5 Molecular Weight (g/mol): 162.14 MDL Number: MFCD00066216 InChI Key: YSEKNCXYRGKTBJ-UHFFFAOYNA-N Synonym: D-(+)-Apple Acid Dimethyl Ester, D-(+)-Malic Acid Dimethyl Ester PubChem CID: 11062697 IUPAC Name: 1,4-dimethyl 2-hydroxybutanedioate SMILES: COC(=O)CC(O)C(=O)OC