missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl D-(+)-Malate 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Quantity | 5 g |
| Formula Weight | 162.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dimethyl D-(+)-Malate |
Sécurité et manipulation
| TSCA | No |