accessibility menu, dialog, popup

UserName

CAS RN 70351-87-0

CH3OOCH3OCH3OH3COH3COCH3

CAS RN 70351-87-0

IUPAC Name: 2,3,6,7,10,11-hexakis(octyloxy)triphenylene
Synonyms: 2,3,6,7,10,11-hexakis(octyloxy)triphenylene
Molecular Weight (g/mol): 997.58
Molecular Formula: C66H108O6
InChi Key: IUFDADHEWRYRSP-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C(OCCCCCCCC)C=C2C(=C1)C1=CC(OCCCCCCCC)=C(OCCCCCCCC)C=C1C1=CC(OCCCCCCCC)=C(OCCCCCCCC)C=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results