accessibility menu, dialog, popup

UserName

CAS RN 703-18-4

IUPAC Name: trifluoromethanesulfinylbenzene
Synonyms: trifluoromethanesulfinylbenzene
Molecular Weight (g/mol): 194.17
Molecular Formula: C7H5F3OS
InChi Key: WZAJOJWOOXXUCT-UHFFFAOYNA-N
SMILES: FC(F)(F)S(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results