missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Trifluoromethyl Sulfoxide 98.0+%, TCI America™

Catalog No. P24605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24605G 5 g
P24601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24605G Supplier TCI America Supplier No. P24605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 703-18-4
Molecular Formula C7H5F3OS
Molecular Weight (g/mol) 194.171
InChI Key WZAJOJWOOXXUCT-UHFFFAOYSA-N
Synonym [(Trifluoromethyl)sulfinyl]benzene
PubChem CID 9794162
IUPAC Name trifluoromethylsulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
Formula Weight 194.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Trifluoromethyl Sulfoxide