accessibility menu, dialog, popup

UserName

ClClN

CAS RN 70258-18-3

IUPAC Name: 2-chloro-5-(chloromethyl)pyridine
Synonyms: 2-chloro-5-(chloromethyl)pyridine
Molecular Weight (g/mol): 162.01
Molecular Formula: C6H5Cl2N
InChi Key: SKCNYHLTRZIINA-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(Cl)N=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)