missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(chloromethyl)pyridine, 96%

Catalog No. AAL1928414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1928414 25 g
AAL1928406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1928414 Supplier Thermo Scientific Chemicals Supplier No. L1928414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70258-18-3 | C6H5Cl2N | 162.013 g/mol

2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 70258-18-3
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00125366
InChI Key SKCNYHLTRZIINA-UHFFFAOYSA-N
Synonym 2-chloro-5-chloromethyl pyridine, pyridine, 2-chloro-5-chloromethyl, 2-chloro-5-chloromethyl-pyridine, pubchem9210, acmc-1bl9y, ksc377c3b, 2-chloro-5chloromethylpyridine, 2-chloro-5-chloromethypyridine, 5-chloromethyl-2-chloropyridine, 6-chloro-3-chloromethylpyridine
PubChem CID 155479
IUPAC Name 2-chloro-5-(chloromethyl)pyridine
SMILES C1=CC(=NC=C1CCl)Cl

Specifications

Melting Point 37°C to 42°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 1635690
Solubility Information Insoluble in water.
Formula Weight 162.02
Percent Purity 96%
Chemical Name or Material 2-Chloro-5-(chloromethyl)pyridine
Hazard Category H302-H314-H334-H335
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only