accessibility menu, dialog, popup

UserName

CH3CH3CH3H3CH3C

CAS RN 700-12-9

IUPAC Name: 1,2,3,4,5-pentamethylbenzene
Synonyms: pentamethylbenzene pentamethylbenzol
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: BEZDDPMMPIDMGJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(C)=C1C
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)