accessibility menu, dialog, popup

UserName

CH3SNH2OO

CAS RN 70-55-3

IUPAC Name: 4-methylbenzene-1-sulfonamide
Synonyms: p-toluenesulfonamide 4-toluenesulfanamide p-toluenesulfonylamide p-tosylamide tosylamide 4-toluenesulfonylamide amide 4-toluenesulfonic acid
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: LMYRWZFENFIFIT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(N)(=O)=O
Boiling Point 
  • (2)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (4)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (2)
  • (6)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)