missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonamide 98.0+%, TCI America™

Catalog No. T0281500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0281500G 500 g
T028125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0281500G Supplier TCI America Supplier No. T0281500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 70-55-3
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00011692
InChI Key LMYRWZFENFIFIT-UHFFFAOYSA-N
Synonym p-toluenesulfonamide, 4-toluenesulfonamide, tosylamide, p-tosylamide, benzenesulfonamide, 4-methyl, p-tolylsulfonamide, toluene-4-sulfonamide, tolylsulfonamide, 4-methylbenzene-1-sulfonamide, p-toluenesulfamide
PubChem CID 6269
ChEBI CHEBI:34435
IUPAC Name 4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 137°C
Color White
Boiling Point 214°C
Quantity 500 g
Formula Weight 171.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonamide
EINECSNumber (3)-1895
RTECSNumber XT5075000
TSCA Yes