accessibility menu, dialog, popup

UserName

CAS RN 70-34-8

IUPAC Name: 1-fluoro-2,4-dinitrobenzene
Synonyms: dinitrofluorobenzene DNFB 1,2,4-fluorodinitrobenzene FDNB DFB fluorodinitrobenzene fluoro-2,4-dinitrobenzene
Molecular Weight (g/mol): 186.1
Molecular Formula: C6H3FN2O4
InChi Key: LOTKRQAVGJMPNV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
Boiling Point 
  • (11)
Molecular Weight (g/mol) 
  • (7)
  • (4)
Percent Purity 
  • (4)
  • (4)
  • (3)
Quantity 
  • (2)
  • (4)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (4)
  • (7)
  • (11)
  • (7)
  • (4)
  • (4)
  • (4)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)