missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrofluorobenzene 99.0+%, TCI America™

Catalog No. A55125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55125G 5 g
A551225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55125G Supplier TCI America Supplier No. A55125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 70-34-8
Molecular Formula C6H3FN2O4
Molecular Weight (g/mol) 186.098
MDL Number MFCD00007056
InChI Key LOTKRQAVGJMPNV-UHFFFAOYSA-N
Synonym 2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
PubChem CID 6264
ChEBI CHEBI:53049
IUPAC Name 1-fluoro-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F

Specifications

Melting Point 26°C
Boiling Point 296°C
Quantity 5 g
UN Number 2811
Formula Weight 186.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dinitrofluorobenzene [for HPLC Labeling]
EINECSNumber (3)-0456
RTECSNumber CZ7800000
TSCA Yes