accessibility menu, dialog, popup

UserName

CAS RN 69999-16-2

IUPAC Name: 2-(2,3-dihydro-1-benzofuran-5-yl)acetic acid
Synonyms: 2,3-dihydro-1-benzofuran-5-ylacetic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: LALSYIKKTXUSLG-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC2=C(OCC2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results