missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrobenzofuran-5-acetic Acid 98.0+%, TCI America™

Catalog No. D42705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42705G 5 g
D42701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42705G Supplier TCI America Supplier No. D42705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69999-16-2
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD01941301
InChI Key LALSYIKKTXUSLG-UHFFFAOYSA-N
Synonym 5-Coumaranacetic Acid
PubChem CID 2737455
IUPAC Name 2-(2,3-dihydro-1-benzofuran-5-yl)acetic acid
SMILES C1COC2=C1C=C(C=C2)CC(=O)O

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydrobenzofuran-5-acetic Acid