accessibility menu, dialog, popup

UserName

H3COOOOCH3OOH3CONNONNH2NH

CAS RN 6979-94-8

IUPAC Name: [3,4-bis(acetyloxy)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-5-(2-amino-6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol): 409.36
Molecular Formula: C16H19N5O8
InChi Key: ULXDFYDZZFYGIY-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(C(OC(C)=O)C1OC(C)=O)N1C=NC2=C1NC(N)=NC2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results