accessibility menu, dialog, popup

UserName

CAS RN 6976-93-8

H3COOOCH3CH2

CAS RN 6976-93-8

IUPAC Name: 2-methoxyethyl 2-methylprop-2-enoate
Synonyms: 2-methoxyethyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: YXYJVFYWCLAXHO-UHFFFAOYSA-N
SMILES: COCCOC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results