missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M2282500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2282500G 500 g
M228225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2282500G Supplier TCI America Supplier No. M2282500G

Chemical Identifiers

CAS 6976-93-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00048122
InChI Key YXYJVFYWCLAXHO-UHFFFAOYSA-N
Synonym Ethylene Glycol Monomethyl Ether Methacrylate, Methacrylic Acid 2-Methoxyethyl Ester
PubChem CID 81466
IUPAC Name 2-methoxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOC

Specifications

Color Colorless
Boiling Point 65°C
Quantity 500 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1049
TSCA Yes