accessibility menu, dialog, popup

UserName

CAS RN 697-86-9

IUPAC Name: 2-bromo-4,6-dichloroaniline
Synonyms: 2-bromo-4,6-dichloroaniline
Molecular Weight (g/mol): 240.91
Molecular Formula: C6H4BrCl2N
InChi Key: DTPADCOGQUOGHT-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Cl)C=C1Br
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)