missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dichloroaniline 97.0+%, TCI America™

Catalog No. B21975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21975G 5 g
B219725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21975G Supplier TCI America Supplier No. B21975G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 697-86-9
Molecular Formula C6H4BrCl2N
Molecular Weight (g/mol) 240.909
MDL Number MFCD00040936
InChI Key DTPADCOGQUOGHT-UHFFFAOYSA-N
Synonym pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline
PubChem CID 2756901
IUPAC Name 2-bromo-4,6-dichloroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Br)Cl

Specifications

Melting Point 81°C
Color White-Yellow
Boiling Point 273°C
Quantity 5 g
Formula Weight 240.91
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,6-dichloroaniline