accessibility menu, dialog, popup

UserName

CAS RN 6968-24-7

IUPAC Name: 2,6-dibromo-4-methylaniline
Synonyms: 2,6-dibromo-4-methylaniline
Molecular Weight (g/mol): 264.95
Molecular Formula: C7H7Br2N
InChi Key: ATDIROHVRVQMRO-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(N)C(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)