accessibility menu, dialog, popup

UserName

CH3H3CNHOOHOHNONH2NHONH2NH2NNH2OHNOHOHO

CAS RN 69558-55-0

IUPAC Name: 3-(6-amino-2-{2-amino-5-[(diaminomethylidene)amino]pentanamido}hexanamido)-3-[(1-{[1-carboxy-2-(4-hydroxyphenyl)ethyl]carbamoyl}-2-methylpropyl)carbamoyl]propanoic acid
Synonyms: 3-(6-amino-2-{2-amino-5-[(diaminomethylidene)amino]pentanamido}hexanamido)-3-[(1-{[1-carboxy-2-(4-hydroxyphenyl)ethyl]carbamoyl}-2-methylpropyl)carbamoyl]propanoic acid
Molecular Weight (g/mol): 679.78
Molecular Formula: C30H49N9O9
InChi Key: PSWFFKRAVBDQEG-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C(CC(O)=O)NC(=O)C(CCCCN)NC(=O)C(N)CCCN=C(N)N)C(=O)NC(CC1=CC=C(O)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)