missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymopentin, 99+%

Catalog No. AAJ61431MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61431MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61431MB Supplier Thermo Scientific Chemicals Supplier No. J61431MB
Only null left

Reduces endocrine responses

Reduces endocrine responses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69558-55-0

Specifications

Purity ≥99%
Color White
Content And Storage Store at -20°C
Format Powder
Molecular Weight (g/mol) 679.776
Product Type Thymopentin
Quantity 25 mg

RUO – Research Use Only