accessibility menu, dialog, popup

UserName

CH3NH2N

CAS RN 695-34-1

IUPAC Name: 4-methylpyridin-2-amine
Synonyms: 2-pyridinamine, 4-methyl-
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: ORLGLBZRQYOWNA-UHFFFAOYSA-N
SMILES: CC1=CC=NC(N)=C1
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)