CAS RN 694-87-1
IUPAC Name:
bicyclo[4.2.0]octa-1,3,5-triene
Synonyms:
benzocyclobutene
Molecular Weight (g/mol):
104.15
Molecular Formula:
C8H8
InChi Key:
UMIVXZPTRXBADB-UHFFFAOYSA-N
SMILES:
C1CC2=CC=CC=C12
1
–
1
of
1
results
1
Benzocyclobutene 94.0+%, TCI America™
CAS: 694-87-1 Molecular Formula: C8H8 Molecular Weight (g/mol): 104.152 MDL Number: MFCD01321219 InChI Key: UMIVXZPTRXBADB-UHFFFAOYSA-N Synonym: Bicyclo[4.2.0]octa-1,3,5-triene PubChem CID: 69667 ChEBI: CHEBI:87328 IUPAC Name: bicyclo[4.2.0]octa-1,3,5-triene SMILES: C1CC2=CC=CC=C21