missing translation for 'onlineSavingsMsg'
Learn More

Benzocyclobutene 94.0+%, TCI America™

Catalog No. B3863200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3863200MG 200 mg
B38631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3863200MG Supplier TCI America Supplier No. B3863200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 694-87-1
Molecular Formula C8H8
Molecular Weight (g/mol) 104.152
MDL Number MFCD01321219
InChI Key UMIVXZPTRXBADB-UHFFFAOYSA-N
Synonym Bicyclo[4.2.0]octa-1,3,5-triene
PubChem CID 69667
ChEBI CHEBI:87328
IUPAC Name bicyclo[4.2.0]octa-1,3,5-triene
SMILES C1CC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 150°C
Quantity 200 mg
UN Number 3295
Formula Weight 104.15
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzocyclobutene