accessibility menu, dialog, popup

UserName

CH3CH3SiOONNOHNOHOOSiOCH3H3CCH3CH3H3CCH3

CAS RN 69304-43-4

IUPAC Name: N-{1-[9-hydroxy-2,2,4,4-tetrakis(propan-2-yl)-hexahydro-2H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-2-oxo-1,2-dihydropyrimidin-4-yl}benzamide
Synonyms: N-(1-{9-hydroxy-2,2,4,4-tetraisopropyl-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl}-2-oxopyrimidin-4-yl)benzamide
Molecular Weight (g/mol): 589.84
Molecular Formula: C28H43N3O7Si2
InChi Key: NFIAUHQTSCWVMS-UHFFFAOYNA-N
SMILES: CC(C)[Si]1(OCC2OC(C(O)C2O[Si](O1)(C(C)C)C(C)C)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results