missing translation for 'onlineSavingsMsg'
Learn More

N4-Benzoyl-3',5'-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine 97.0+%, TCI America™

Catalog No. B36315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36315G 5 g
B36311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36315G Supplier TCI America Supplier No. B36315G

Chemical Identifiers

CAS 69304-43-4
Molecular Formula C28H43N3O7Si2
Molecular Weight (g/mol) 589.836
InChI Key NFIAUHQTSCWVMS-LYPBTDJXSA-N
PubChem CID 10054127
IUPAC Name N-[1-[(6aR,8R,9R,9aS)-9-hydroxy-2,2,4,4-tetra(propan-2-yl)-6a,8,9,9a-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-2-oxopyrimidin-4-yl]benzamide
SMILES CC(C)[Si]1(OCC2C(C(C(O2)N3C=CC(=NC3=O)NC(=O)C4=CC=CC=C4)O)O[Si](O1)(C(C)C)C(C)C)C(C)C

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 589.84
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N4-Benzoyl-3′,5′-O-(1,1,3,3-tetraisopropyl-1,3-disiloxanediyl)cytidine