accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 693-38-9

IUPAC Name: ethenyl hexadecanoate
Synonyms: ethenyl hexadecanoate
Molecular Weight (g/mol): 282.47
Molecular Formula: C18H34O2
InChi Key: UJRIYYLGNDXVTA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results