accessibility menu, dialog, popup

UserName

CAS RN 6914-71-2

IUPAC Name: 1,1-dimethyl cyclopropane-1,1-dicarboxylate
Synonyms: 1,1-dimethyl cyclopropane-1,1-dicarboxylate
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: PWLLZZMFFZUSOG-UHFFFAOYSA-N
SMILES: COC(=O)C1(CC1)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results