missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,1-Cyclopropanedicarboxylate 98.0+%, TCI America™

Catalog No. C1471500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1471500G 500 g
C147125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1471500G Supplier TCI America Supplier No. C1471500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6914-71-2
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.153
MDL Number MFCD00192079
InChI Key PWLLZZMFFZUSOG-UHFFFAOYSA-N
Synonym 1,1-Cyclopropanedicarboxylic Acid Dimethyl Ester
PubChem CID 2769539
IUPAC Name dimethyl cyclopropane-1,1-dicarboxylate
SMILES COC(=O)C1(CC1)C(=O)OC

Specifications

Color Yellow
Boiling Point 198°C
Quantity 500 g
Formula Weight 158.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 1,1-Cyclopropanedicarboxylate