accessibility menu, dialog, popup

UserName

CAS RN 69075-47-4

CHNONNH2OOHOH

CAS RN 69075-47-4

IUPAC Name: 4-amino-5-ethynyl-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: 4-amino-5-ethynyl-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Molecular Weight (g/mol): 251.24
Molecular Formula: C11H13N3O4
InChi Key: HMJSYXIPNSASJY-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1C#C)C1CC(O)C(CO)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results