missing translation for 'onlineSavingsMsg'
Learn More

5-Ethynyl-2'-deoxycytidine 98.0+%, TCI America™

Catalog No. E1093200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1093200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E1093200MG Supplier TCI America Supplier No. E1093200MG
Only null left

Chemical Identifiers

CAS 69075-47-4
Molecular Formula C11H13N3O4
Molecular Weight (g/mol) 251.242
MDL Number MFCD22417249
InChI Key HMJSYXIPNSASJY-IWSPIJDZSA-N
Synonym EdC
PubChem CID 90176383
IUPAC Name 4-amino-5-ethynyl-1-[(2R,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C#CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O

Specifications

Melting Point 190°C
Quantity 200 mg
Formula Weight 251.24
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Ethynyl-2′-deoxycytidine