accessibility menu, dialog, popup

UserName

CH3OOO(R)OH(S)(S)OH

CAS RN 6902-77-8

IUPAC Name: methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1H,4aH,5H,7aH-cyclopenta[c]pyran-4-carboxylate
Synonyms: genipin
Molecular Weight (g/mol): 226.23
Molecular Formula: C11H14O5
InChi Key: AZKVWQKMDGGDSV-BCMRRPTOSA-N
SMILES: COC(=O)C1=CO[C@@H](O)[C@H]2[C@@H]1CC=C2CO
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results