accessibility menu, dialog, popup

UserName

OH(R)HS(S)OHSH

CAS RN 6892-68-8

IUPAC Name: (2R,3S)-1,4-disulfanylbutane-2,3-diol
Synonyms: dithioerythritol
Molecular Weight (g/mol): 154.24
Molecular Formula: C4H10O2S2
InChi Key: VHJLVAABSRFDPM-ZXZARUISSA-N
SMILES: O[C@@H](CS)[C@H](O)CS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results