missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Dithioerythritol, Calbiochem™,

Catalog No. 2331525GM
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2331525GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2331525GM Supplier MilliporeSigma™ Supplier No. 2331525GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6892-68-8
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00063750
InChI Key VHJLVAABSRFDPM-ZXZARUISSA-N
Synonym dithioerythritol, 1,4-dithioerythritol, dte, 2r,3s-1,4-dimercaptobutane-2,3-diol, erythro-1,4-dimercapto-2,3-butanediol, 2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel, 1,4-dithioerythritol dte, 2r*,3s*-1,4-dimercapto-2,3-butanediol, erythro-2,3-dihydroxy-1,4-butanedithiol, 2r,3s-1,4-disulfanylbutane-2,3-diol
PubChem CID 439352
ChEBI CHEBI:17456
IUPAC Name (2R,3S)-1,4-disulfanylbutane-2,3-diol
SMILES O[C@@H](CS)[C@H](O)CS

Specifications

Color White
Packaging Glass Bottle
Quantity 5 g