accessibility menu, dialog, popup

UserName

CAS RN 68892-41-1

IUPAC Name: N-{9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide
Synonyms: N-{9-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl}-2-methylpropanamide
Molecular Weight (g/mol): 639.71
Molecular Formula: C35H37N5O7
InChi Key: RMQXDNUKLIDXOS-ZGIBFIJWSA-N
SMILES: COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1NC(NC(=O)C(C)C)=NC2=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results