accessibility menu, dialog, popup

UserName

CH3CH3CH3OOOOOCH3H3CH3C

CAS RN 68835-89-2

IUPAC Name: bis(2-methylbutan-2-yl) dicarbonate
Synonyms: bis(2-methylbutan-2-yl) dicarbonate
Molecular Weight (g/mol): 246.3
Molecular Formula: C12H22O5
InChi Key: PGWKKYVDHYLHNF-UHFFFAOYSA-N
SMILES: CCC(C)(C)OC(=O)OC(=O)OC(C)(C)CC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results