accessibility menu, dialog, popup

UserName

CAS RN 68282-53-1

IUPAC Name: 5-methyl-1H-imidazole-4-carbaldehyde
Synonyms: 5-methyl-1H-imidazole-4-carbaldehyde
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: KMWCSNCNHSEXIF-UHFFFAOYSA-N
SMILES: CC1=C(C=O)N=CN1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)