accessibility menu, dialog, popup

UserName

CH3NH3CCH3FFF

CAS RN 68254-41-1

IUPAC Name: N,N,N-trimethyl-3-(trifluoromethyl)anilinium
Synonyms: N,N,N-trimethyl-3-(trifluoromethyl)anilinium
Molecular Weight (g/mol): 204.22
Molecular Formula: C10H13F3N
InChi Key: ZLBCBMSHZLXLQF-UHFFFAOYSA-N
SMILES: C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results