missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyltrimethylammonium Hydroxide (5% in Methanol), TCI America™

Catalog No. T096125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T096125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T096125ML Supplier TCI America Supplier No. T096125ML

[for Transesterification of Glyceride]

Chemical Identifiers

CAS 68254-41-1
Molecular Formula C10H14F3NO
Molecular Weight (g/mol) 221.223
MDL Number MFCD00059473
InChI Key BFPOZPZYPNVMHU-UHFFFAOYSA-M
Synonym 3-trifluoromethyl phenyltrimethylammonium hydroxide, m-tfptah, trifluoromethylphenyltrimethylammoniumhydroxide, 3-trifluoromethyl phenyltrimethylammonium hydroxide in methanol, n,n,n-trimethyl-3-trifluoromethyl anilinium hydroxide, trimethyl-3-trifluoromethyl phenyl azanium hydroxide, m-trifluoromethylphenyl trimethylammonium hydroxide, trimethyl-3-trifluoromethylphenylammonium hydroxide, n,n,n-trimethyl-3-trifluoromethyl anilinium oxidanide, trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium hydroxide
PubChem CID 2760772
IUPAC Name trimethyl-[3-(trifluoromethyl)phenyl]azanium;hydroxide
SMILES C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[OH-]

Specifications

Color Colorless
Quantity 25 mL
UN Number 2924
Formula Weight 221.22
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)phenyltrimethylammonium Hydroxide (5% in Methanol) [for Transesterification of Gl
TSCA No
Recommended Storage Refrigerator