accessibility menu, dialog, popup

UserName

CAS RN 67816-09-5

LaH3CCH3OOH3CCH3OOH3CCH3OO

CAS RN 67816-09-5

IUPAC Name: lanthanum(3+) tris(2-ethylhexanoate)
Synonyms: lanthanum(3+) tris(2-ethylhexanoate)
Molecular Weight (g/mol): 568.52
Molecular Formula: C24H45LaO6
InChi Key: PPNFILUQDVDXDA-UHFFFAOYNA-K
SMILES: [La+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results