CAS RN 67494-15-9
IUPAC Name:
(4aR,10aS)-6-hydroxy-1,1-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,9,10,10a-octahydrophenanthrene-4a-carboxylic acid
Synonyms:
pisiferic acid
Molecular Weight (g/mol):
316.44
Molecular Formula:
C20H28O3
InChi Key:
ATHWSPHADLLZSS-PXNSSMCTSA-N
SMILES:
CC(C)C1=CC2=C(C=C1O)[C@]1(CCCC(C)(C)[C@@H]1CC2)C(O)=O
1
–
1
of
1
results
1
Pisiferic Acid 98.0+%, TCI America™
CAS: 67494-15-9 Molecular Formula: C20H28O3 Molecular Weight (g/mol): 316.441 MDL Number: MFCD01708794 InChI Key: ATHWSPHADLLZSS-PXNSSMCTSA-N PubChem CID: 162209 ChEBI: CHEBI:70576 IUPAC Name: (4aR,10aS)-6-hydroxy-1,1-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-4a-carboxylic acid SMILES: CC(C)C1=C(C=C2C(=C1)CCC3C2(CCCC3(C)C)C(=O)O)O