missing translation for 'onlineSavingsMsg'
Learn More

Pisiferic Acid 98.0+%, TCI America™

Catalog No. P1933100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1933100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1933100MG Supplier TCI America Supplier No. P1933100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67494-15-9
Molecular Formula C20H28O3
Molecular Weight (g/mol) 316.441
MDL Number MFCD01708794
InChI Key ATHWSPHADLLZSS-PXNSSMCTSA-N
PubChem CID 162209
ChEBI CHEBI:70576
IUPAC Name (4aR,10aS)-6-hydroxy-1,1-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-4a-carboxylic acid
SMILES CC(C)C1=C(C=C2C(=C1)CCC3C2(CCCC3(C)C)C(=O)O)O

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 100 mg
Formula Weight 316.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pisiferic Acid
RTECSNumber SF7325030
TSCA No