accessibility menu, dialog, popup

UserName

CAS RN 67483-13-0

IUPAC Name: disodium 5-isothiocyanato-2-[(1E)-2-(4-isothiocyanato-2-sulfonatophenyl)ethenyl]benzene-1-sulfonate
Synonyms: disodium 5-isothiocyanato-2-[(1E)-2-(4-isothiocyanato-2-sulfonatophenyl)ethenyl]benzenesulfonate
Molecular Weight (g/mol): 498.47
Molecular Formula: C16H8N2Na2O6S4
InChi Key: GEPAYBXVXXBSKP-SEPHDYHBSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1\C=C\C1=CC=C(C=C1S([O-])(=O)=O)N=C=S)N=C=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results