missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4,4'-Diisothiocyanato-2,2'-stilbenedisulfonate Hydrate 90.0+%, TCI America™

Catalog No. D2475250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2475250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2475250MG Supplier TCI America Supplier No. D2475250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67483-13-0
Molecular Formula C16H8N2Na2O6S4
Molecular Weight (g/mol) 498.468
MDL Number MFCD00009638
InChI Key GEPAYBXVXXBSKP-SEPHDYHBSA-L
Synonym dids, unii-1vfrq7416a, disodium 4,4'-diisothiocyanato-2,2'-stilbenedisulfonate, 4,4'-diisothiocyanato-2,2'-stilbenedisulfonic acid disodium salt, 4,4'-diisothiocyano-2,2'-stilbenedisulfonic acid, disodium salt, disodium 5-isothiocyanato-2-e-2-4-isothiocyanato-2-sulfonatophenyl ethenyl benzenesulfonate, spectrum1505164, 4 4'-diisothiocyanato-2 2'-stilbenedisu, disodium 4,4'-diisothiocyanatostilbene-2,2'-disulfonate, disodium 4,4'-diisothiocyanato-2,2'-stilbenedisulfonate, e
PubChem CID 5702690
IUPAC Name disodium;5-isothiocyanato-2-[(E)-2-(4-isothiocyanato-2-sulfonatophenyl)ethenyl]benzenesulfonate
SMILES C1=CC(=C(C=C1N=C=S)S(=O)(=O)[O-])C=CC2=C(C=C(C=C2)N=C=S)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 250 mg
Formula Weight 498.47
Percent Purity ≥90.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4,4′-Diisothiocyanato-2,2′-stilbenedisulfonate Hydrate
TSCA No
Recommended Storage Refrigerator